Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: BRD469, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23456501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23456501

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity All
Isotype IgG1
Concentration 1mg/mL
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Quantity 0.1mg
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Format Purified
Reconstitution Protein A purified
Regulatory Status RUO
Conjugate Unlabeled
Purification Method Purified
Clone BRD469
Monoclonal or Polyclonal Monoclonal
Antigen Bromodeoxyuridine/BrdU
Formulation 10mM PBS. Does not contain BSA. Azide Free.