Please login to your online account to display your discounted pricing

Mouse anti-Biotin (Vitamin B7 or Vitamin H), Clone: SPM375, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Clone SPM375
Concentration 0.2mg/mL
Immunogen Biotinylated sheep immunoglobulin
Formulation PBS with 0.05% BSA. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23281201

 
novus biologicals canada
NBP23281201ML
0.1mL Each for $139.00

NBP23281210

 
novus biologicals canada
NBP23281210ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Clone SPM375
Concentration 0.2mg/mL
Immunogen Biotinylated sheep immunoglobulin
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity All
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting