Please login to your online account to display your discounted pricing

Mouse anti-Biotin (Vitamin B7 or Vitamin H), Clone: SPM375, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23281201ML

 View all options for this product


For Research Use Only

Catalog No. NBP23281201


Description & Specifications


Antigen Biotin (Vitamin B7 or Vitamin H)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM375
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity All
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal