Please login to your online account to display your discounted pricing

Mouse anti-Biotin (Vitamin B7 or Vitamin H), Clone: SPM375, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23281210ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23281210

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Biotinylated sheep immunoglobulin
Formulation PBS with 0.05% BSA. Protein G purified.
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM375
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity All
Antigen Biotin (Vitamin B7 or Vitamin H)
Isotype IgG1