Please login to your online account to display your discounted pricing

Mouse anti-Biotin (Vitamin B7 or Vitamin H), Clone: SPM375, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Clone SPM375
Concentration 1mg/mL
Immunogen Biotinylated sheep immunoglobulin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23477101

 
novus biologicals canada
NBP23477101MG
0.1mg Each for $384.00

NBP23477102

 
novus biologicals canada
NBP23477102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Biotin (Vitamin B7 or Vitamin H)
Clone SPM375
Concentration 1mg/mL
Immunogen Biotinylated sheep immunoglobulin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot