Please login to your online account to display your discounted pricing

Mouse anti-Biotin (Vitamin B7 or Vitamin H), Clone: SPM375, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity All
Antigen Biotin (Vitamin B7 or Vitamin H)
Format Purified
Host Species Mouse
Immunogen Biotinylated sheep immunoglobulin
Isotype IgG1
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone SPM375
Concentration 1mg/mL
Conjugate Unlabeled