Please login to your online account to display your discounted pricing

Mouse anti-Biotin (Vitamin B7 or Vitamin H), Clone: Hyb-8, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464601

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Immunogen Biotinylated sheep immunoglobulin
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Cross Reactivity All
Antigen Biotin (Vitamin B7 or Vitamin H)
Host Species Mouse
Isotype IgG1
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Clone Hyb-8
Reconstitution Protein A purified
Regulatory Status RUO
Purification Method Purified
Conjugate Unlabeled
Primary or Secondary Primary