Please login to your online account to display your discounted pricing

Mouse anti-beta Tubulin, Clone: 10B1, Novus Biologicals™

$278.00 - $354.00

Specifications

Antigen beta Tubulin
Clone 10B1
Concentration 1mg/mL
Immunogen The immunogen for this antibody is produced against beta-Tubulin.
Gene ID (Entrez) 203068
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14138601

 
novus biologicals canada
NBP14138601ML
0.1mL Each for $278.00

NBP14138602

 
novus biologicals canada
NBP14138602ML
0.2mL Each for $354.00
Description & Specifications

Specifications

Antigen beta Tubulin
Clone 10B1
Concentration 1mg/mL
Immunogen The immunogen for this antibody is produced against beta-Tubulin.
Gene ID (Entrez) 203068
Formulation PBS
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting