Please login to your online account to display your discounted pricing

Mouse anti-beta Tubulin, Clone: 10B1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14138602ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14138602

  • / Each
 

Description & Specifications

Specifications

Antigen beta Tubulin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Clone 10B1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS
Host Species Mouse
Immunogen The immunogen for this antibody is produced against beta-Tubulin.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.2mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 203068