Please login to your online account to display your discounted pricing

Mouse anti-beta Tubulin, Clone: 10B1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14138601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14138601

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Immunogen The immunogen for this antibody is produced against beta-Tubulin.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Affinity Purified
Isotype IgG1
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Clone 10B1
Formulation PBS
Quantity 0.1mL
Gene ID (Entrez) 203068
Conjugate Unlabeled
Antigen beta Tubulin
Concentration 1mg/mL