Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: SPM129, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Clone SPM129
Immunogen Human peripheral blood mononuclear cells
Gene Accession No. Q9P2W7
Gene ID (Entrez) 27087
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23444901

 
novus biologicals canada
NBP23444901ML
0.1mL Each for $139.00

NBP23444905

 
novus biologicals canada
NBP23444905ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Clone SPM129
Immunogen Human peripheral blood mononuclear cells
Gene Accession No. Q9P2W7
Gene ID (Entrez) 27087
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence