Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: SPM129, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444901

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Immunogen Human peripheral blood mononuclear cells
Primary or Secondary Primary
Cross Reactivity Human
Formulation Culture Medium. Bioreactor concentrate.
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 27087
Test Specificity Does not react to Rat.
Clone SPM129
Conjugate Unlabeled
Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Format Concentrated
Gene Accession No. Q9P2W7
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.