Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: SPM129, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444905

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Immunogen Human peripheral blood mononuclear cells
Cross Reactivity Human
Formulation Culture Medium. Bioreactor concentrate.
Test Specificity Does not react to Rat.
Host Species Mouse
Gene Accession No. Q9P2W7
Quantity 0.5mL
Gene ID (Entrez) 27087
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Isotype IgM, κ
Format Concentrated
Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Clone SPM129
Purification Method Concentrated
Regulatory Status RUO
Conjugate Unlabeled