Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: NK-1, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Clone NK-1
Immunogen Human peripheral blood mononuclear cells were used as immunogen.
Gene Accession No. Q96E93
Gene ID (Entrez) 27087
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21519205

 
novus biologicals canada
NBP21519205ML
0.5mL Each for $321.00

NBP21519201

 
novus biologicals canada
NBP21519201ML
0.1mL Each for $139.00

NBP21519210

 
novus biologicals canada
NBP21519210ML
1mL Each for $429.00
Description & Specifications

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Clone NK-1
Immunogen Human peripheral blood mononuclear cells were used as immunogen.
Gene Accession No. Q96E93
Gene ID (Entrez) 27087
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting