Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: NK-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21519205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21519205

  • / Each
 

Description & Specifications

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Format Concentrated
Gene Accession No. Q96E93
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.5mL
Regulatory Status RUO
Test Specificity Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.
Cross Reactivity Human
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Isotype IgM, κ
Purification Method Concentrated
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 27087
Clone NK-1
Conjugate Unlabeled
Immunogen Human peripheral blood mononuclear cells were used as immunogen.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.