Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: NK-1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP21519210ML

 View all options for this product


For Research Use Only

Catalog No. NBP21519210


Description & Specifications


Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone NK-1
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. Q96E93
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells were used as immunogen.
Isotype IgM, κ
Purification Method Concentrated
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 27087
Test Specificity Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.