Please login to your online account to display your discounted pricing

Mouse anti-beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57, Clone: NK-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21519201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21519201

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Human peripheral blood mononuclear cells were used as immunogen.
Regulatory Status RUO
Host Species Mouse
Isotype IgM, κ
Conjugate Unlabeled
Test Specificity Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.
Gene Accession No. Q96E93
Clone NK-1
Purification Method Concentrated
Cross Reactivity Human
Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1/CD57
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 27087
Formulation Culture Medium. Bioreactor concentrate.
Format Concentrated
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mL