Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: SPM165, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Bcl-xL
Clone SPM165
Concentration 0.2mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23281001

 
novus biologicals canada
NBP23281001ML
0.1mL Each for $139.00

NBP23281010

 
novus biologicals canada
NBP23281010ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Bcl-xL
Clone SPM165
Concentration 0.2mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
Gene ID (Entrez) 598
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting