Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: SPM165, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23281001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23281001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Concentration 0.2mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen A synthetic peptide
Isotype IgG2a
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Conjugate Unlabeled
Antigen Bcl-xL
Gene Accession No. Q07817
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 598
Clone SPM165