Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: SPM165, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23281010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23281010

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-xL
Format Purified
Gene Accession No. Q07817
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Quantity 1.0mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Formulation PBS with 0.05% BSA.
Host Species Mouse
Isotype IgG2a
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 598
Reconstitution Protein A purified
Clone SPM165
Concentration 0.2mg/mL