missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: SPM165, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Bcl-xL
Clone SPM165
Concentration 1mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23474301 Novus Biologicals Canada
NBP23474301MG
0.1mg Each for $384.00
NBP23474302 Novus Biologicals Canada
NBP23474302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Bcl-xL
1mg/mL
Q07817
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM165
A synthetic peptide
598
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot