Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: SPM165, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474302

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-xL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q07817
Host Species Mouse
Immunogen A synthetic peptide
Isotype IgG2a
Purification Method Purified
Quantity 0.2mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 598
Test Specificity Recognizes a protein of 27kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2a or Bax protein.
Reconstitution Protein A purified
Clone SPM165
Concentration 1mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.