Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: SPM165, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474301

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-xL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM165
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q07817
Host Species Murine
Immunogen A synthetic peptide
Isotype IgG2a
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 598
Test Specificity Recognizes a protein of 27kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2a or Bax protein.
Reconstitution Protein A purified
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rat