Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: BX006 + 2H12, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Bcl-xL
Clone BX006 + 2H12
Concentration 0.2mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23429001

 
novus biologicals canada
NBP23429001ML
0.1mL Each for $139.00

NBP23429005

 
novus biologicals canada
NBP23429005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Bcl-xL
Clone BX006 + 2H12
Concentration 0.2mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
Gene ID (Entrez) 598
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting