Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: BX006 + 2H12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457101

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Quantity 0.1mg
Regulatory Status RUO
Host Species Mouse
Gene ID (Entrez) 598
Purification Method Purified
Gene Accession No. Q07817
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Immunogen A synthetic peptide
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Antigen Bcl-xL
Reconstitution Protein A purified
Clone BX006 + 2H12
Monoclonal or Polyclonal Monoclonal