Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 7D9, Novus Biologicals™

$307.00 - $518.93

Specifications

Antigen Bcl-xL
Clone 7D9
Immunogen Synthetic peptide corresponding to N-terminal of mouse Bcl-xL protein.
Gene ID (Entrez) 598
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14976101

 
novus biologicals canada
NBP14976101ML
0.1mL Each for $307.00

NBP14976105

 
novus biologicals canada
NBP14976105ML
0.5mL Each for $363.90

NBP14976110

 
novus biologicals canada
NBP14976110ML
1mL Each for $518.93
Description & Specifications

Specifications

Antigen Bcl-xL
Clone 7D9
Immunogen Synthetic peptide corresponding to N-terminal of mouse Bcl-xL protein.
Gene ID (Entrez) 598
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2a
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)