Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 7D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14976105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14976105

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Bcl-xL
Format Affinity Purified
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Immunogen Synthetic peptide corresponding to N-terminal of mouse Bcl-xL protein.
Isotype IgG2a
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 598
Clone 7D9
Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Purification Method Affinity Purified
Regulatory Status RUO