Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 7D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14976170ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14976170

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 7mL
Conjugate Unlabeled
Immunogen Synthetic peptide corresponding to N-terminal of mouse Bcl-xL protein.
Storage Requirements Store at 4°C. Do not freeze.
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Isotype IgG2a
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Antigen Bcl-xL
Gene ID (Entrez) 598
Regulatory Status RUO
Purification Method Affinity Purified
Clone 7D9
Host Species Mouse
Primary or Secondary Primary