Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 7D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14976110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14976110

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Isotype IgG2a
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Host Species Mouse
Primary or Secondary Primary
Immunogen Synthetic peptide corresponding to N-terminal of mouse Bcl-xL protein.
Purification Method Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Antigen Bcl-xL
Regulatory Status RUO
Quantity 1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 598
Clone 7D9