Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 7D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14976101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14976101

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide corresponding to N-terminal of mouse Bcl-xL protein.
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Bcl-xL
Format Affinity Purified
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Host Species Mouse
Isotype IgG2a
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 598
Clone 7D9
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.