Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 2H12, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Bcl-xL
Clone 2H12
Concentration 1mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23453101

 
novus biologicals canada
NBP23453101MG
0.1mg Each for $384.00

NBP23453102

 
novus biologicals canada
NBP23453102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Bcl-xL
Clone 2H12
Concentration 1mg/mL
Immunogen A synthetic peptide
Gene Accession No. Q07817
Gene ID (Entrez) 598
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG2a
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting