Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 2H12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Clone 2H12
Quantity 0.1mg
Primary or Secondary Primary
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Bcl-xL
Regulatory Status RUO
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Purification Method Purified
Isotype IgG2a
Format Purified
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q07817
Gene ID (Entrez) 598
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.