Please login to your online account to display your discounted pricing

Mouse anti-Bcl-xL, Clone: 2H12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453102

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 598
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Antigen Bcl-xL
Primary or Secondary Primary
Format Purified
Immunogen A synthetic peptide
Quantity 0.2mg
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Clone 2H12
Conjugate Unlabeled
Host Species Mouse
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG2a
Concentration 1mg/mL
Reconstitution Protein A purified
Gene Accession No. Q07817