Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM530, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23438905ML

 View all options for this product


For Research Use Only

Catalog No. NBP23438905


Description & Specifications


Antigen Bcl-2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM530
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10415
Host Species Mouse
Immunogen A synthetic peptide
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 596
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.
Reconstitution Protein A purified