Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM530, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Antigen Bcl-2
Primary or Secondary Primary
Concentration 0.2mg/mL
Cross Reactivity Human
Format Purified
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Immunogen A synthetic peptide
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.
Isotype IgG1, κ
Quantity 0.1mL
Host Species Mouse
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Clone SPM530
Gene ID (Entrez) 596
Gene Accession No. P10415
Reconstitution Protein A purified