Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM530, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen Bcl-2
Format Purified
Host Species Mouse
Immunogen A synthetic peptide
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.
Reconstitution Protein A purified
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P10415
Purification Method Purified
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 596
Clone SPM530
Concentration 1mg/mL