Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Quantity 0.1mL
Cross Reactivity Human
Antigen Bcl-2
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen A synthetic peptide
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P10415
Gene ID (Entrez) 596
Reconstitution Protein A purified
Clone SPM117