Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439105

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-2
Format Purified
Gene Accession No. P10415
Host Species Mouse
Immunogen A synthetic peptide
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 596
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.
Reconstitution Protein A purified
Clone SPM117
Concentration 0.2mg/mL