Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM117, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444002

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Immunogen A synthetic peptide
Isotype IgG1, κ
Regulatory Status RUO
Clone SPM117
Antigen Bcl-2
Gene ID (Entrez) 596
Primary or Secondary Primary
Format Purified
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P10415
Purification Method Purified
Reconstitution Protein A purified
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.