Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: SPM117, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444001

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide
Antigen Bcl-2
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Quantity 0.1mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Clone SPM117
Test Specificity This antibody recognizes a protein of 25-26kDa, identified as the bcl-2alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Does not react with Mouse or Rat.
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Format Purified
Gene ID (Entrez) 596
Purification Method Purified
Primary or Secondary Primary
Regulatory Status RUO
Concentration 1mg/mL
Gene Accession No. P10415
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Host Species Mouse