Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 124, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Immunogen A synthetic peptide
Isotype IgG1, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen Bcl-2
Format Purified
Gene Accession No. P10415
Host Species Mouse
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 596
Test Specificity Does not react to Mouse or Rat.
Reconstitution Protein A purified
Clone 124
Concentration 0.2mg/mL
Conjugate Unlabeled