Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 124, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444305

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-2
Format Purified
Gene Accession No. P10415
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Isotype IgG1, κ
Quantity 0.5mL
Reconstitution Protein A purified
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 596
Test Specificity Does not react to Mouse or Rat.
Clone 124
Concentration 0.2mg/mL
Cross Reactivity Human