Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 124, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Primary or Secondary Primary
Immunogen A synthetic peptide
Quantity 0.1mg
Gene ID (Entrez) 596
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P10415
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Bcl-2
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Reconstitution Protein A purified
Concentration 1mg/mL
Clone 124
Host Species Mouse
Test Specificity Does not react to Mouse or Rat.
Formulation 10mM PBS. Does not contain BSA. Azide Free.