Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 100/D5, Novus Biologicals™

$329.00 - $472.00

Specifications

Antigen Bcl-2
Clone 100/D5
Concentration 0.2mg/mL
Immunogen 1990).
Gene Accession No. P10415
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21520001

 
novus biologicals canada
NBP21520001MG
0.1mg Each for $329.00

NBP21520002

 
novus biologicals canada
NBP21520002MG
0.2mg Each for $472.00
Description & Specifications

Specifications

Antigen Bcl-2
Clone 100/D5
Concentration 0.2mg/mL
Immunogen 1990).
Gene Accession No. P10415
Gene ID (Entrez) 596
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot