Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 100/D5+124, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23444401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23444401

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 596
Gene Accession No. P10415
Conjugate Unlabeled
Format Purified
Isotype IgG1, κ
Immunogen A synthetic peptide
Quantity 0.1mL
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation PBS with 0.05% BSA.
Clone 100/D5+124
Concentration 0.2mg/mL
Test Specificity Does not react to Mouse or Rat.
Reconstitution Protein A purified
Antigen Bcl-2