missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 100/D5+124, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Bcl-2
Clone 100/D5+124
Concentration 1mg/mL
Immunogen A synthetic peptide
Gene Accession No. P10415
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23451501 Novus Biologicals Canada
NBP23451501MG
0.1mg Each for $384.00
NBP23451502 Novus Biologicals Canada
NBP23451502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Bcl-2
1mg/mL
P10415
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
100/D5+124
A synthetic peptide
596
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot