Please login to your online account to display your discounted pricing

Mouse anti-Bcl-2, Clone: 100/D5+124, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Antigen Bcl-2
Gene Accession No. P10415
Test Specificity Does not react to Mouse or Rat.
Quantity 0.1mg
Host Species Mouse
Isotype IgG1, κ
Immunogen A synthetic peptide
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone 100/D5+124
Purification Method Purified
Format Purified
Gene ID (Entrez) 596
Concentration 1mg/mL