Please login to your online account to display your discounted pricing

Mouse anti-Bcl-10, Clone: SPM520, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23436801ML

 View all options for this product


For Research Use Only

Catalog No. NBP23436801


Description & Specifications


Antigen Bcl-10
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM520
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. O95999
Host Species Mouse
Immunogen Human BCL10 recombinant protein (epitope aa122-168)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8915
Test Specificity This MAb labels subpopulations of normal B and T cells and is a useful tool for the sub-classification of lymphomas.
Reconstitution Protein A purified