missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Bcl-10, Clone: SPM520, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Bcl-10
Clone SPM520
Concentration 1mg/mL
Immunogen Human BCL10 recombinant protein (epitope aa122-168)
Gene Accession No. O95999
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23441501 Novus Biologicals Canada
NBP23441501MG
0.1mg Each for $384.00
NBP23441502 Novus Biologicals Canada
NBP23441502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Bcl-10
1mg/mL
O95999
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM520
Human BCL10 recombinant protein (epitope aa122-168)
8915
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot