Please login to your online account to display your discounted pricing

Mouse anti-Bcl-10, Clone: SPM520, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441502

  • / Each
 

Description & Specifications

Specifications

Purification Method Purified
Quantity 0.2mg
Concentration 1mg/mL
Cross Reactivity Human
Antigen Bcl-10
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human BCL10 recombinant protein (epitope aa122-168)
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8915
Test Specificity This MAb labels subpopulations of normal B and T cells and is a useful tool for the sub-classification of lymphomas.
Reconstitution Protein A purified
Clone SPM520
Conjugate Unlabeled
Gene Accession No. O95999